Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553246
Description
Immunogen sequence: YEGIKVGKAK LLNQVLVNLP TSISQQTIAE TDGDSAGSDS FPVGRTHHRW YLC Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 62%.
Copper is a trace element that plays an essential role for organisms that utilize oxygen during respiration. CTR2 is a membrane protein that is a member of the SLC31A transporter family. Although the function of CTR2 is still poorly understood in mammalian cells, it is expected to be involved in low-affinity copper uptake. It is believed to be confined to lysosomes which stimulate copper delivery to the the cytosol of human cells at high concentrations.
Specifications
CTR2 | |
Polyclonal | |
Unconjugated | |
SLC31A2 | |
AI604396; Copper transporter 2; COPT2; CTR2; hCTR2; probable low affinity copper uptake protein 2; RP11-9H12.2; SLC31A2; solute carrier family 31 (copper transporter), member 2; solute carrier family 31 (copper transporters), member 2; solute carrier family 31 member 2; solute carrier family 31, member 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1318 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15432 | |
SLC31A2 | |
Recombinant protein corresponding to Human CTR2. Recombinant protein control fragment (Product #RP-91104). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction