Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTR9 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen CTR9
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095901
View Documents
Novus Biologicals
NBP304380100UL
100 μg Each for $320.71
Add to cart
 
NB095902
View Documents
Novus Biologicals
NBP30438020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

CTR9 Polyclonal antibody specifically detects CTR9 in Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

CTR9
Unconjugated
Rabbit
Chromatin Research
PBS with 50% glycerol, pH7.3.
9646
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry
Purified
RUO
Rat
Ctr9, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), KIAA0155SH2 domain-binding protein 1, p150TSP, RNA polymerase-associated protein CTR9 homolog, SH2 domain binding protein 1 (tetratricopeptide repeat containing), SH2BP1p150, TPR-containing, SH2-binding phosphoprotein, TSBP
Recombinant fusion protein containing a sequence corresponding to amino acids 1024-1173 of human CTR9 (NP_055448.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit