Learn More
Associates of Cape Cod Inc CTRL STD ENDO TOXIN 0.5MG
SDP
Supplier:  Associates of Cape Cod Inc E00051
Encompass
CONTROL STANDARD ENDOTOXIN (CSE), 0.5ug/vial, SINGLE VIAL, reconstitue with LRW ONLY
Catalog No. NC0251085
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.