Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTRP7, Mouse anti-Human,Mouse, Janelia Fluor 646, Clone: Anna-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP197528JF646

Catalog No. NB004663

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CTRP7 Monoclonal antibody specifically detects CTRP7 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

CTRP7
Anna-1
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified, Conjugated
C1QTNF7
Recombinant mouse CTRP7 (C1q tumor necrosis factor-alpha-related protein 7) (aa 18-290).
Protein G purified
RUO
Primary
Human, Mouse
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit