Learn More
Gibco™ CTS™ KnockOut™ SR XenoFree Kit Non-distribution product as customer accommodation.
CTS™ KnockOut™ SR XenoFree Kit
Manufacturer:  Gibco™ A1099201
 View more versions of this product
Catalog No. A1099201
Add to cart
This item is not returnable. View return policy
Description
Description
GIBCO™ KnockOut™ SR XenoFree CTS™ enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins, which facilitates the transition of hESC and hiPSC research from the bench to the clinic.
The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
  • Defined, xenofree medium for hESC⁄iPSC stem cell culture.
  • First-to-market xenofree hESC⁄iPSC product intended for Ex Vivo tissue and cell culture processing applications.
    GIBCO™ KnockOut™ SR XenoFree CTS™ does not contain bovine or other non-human, animal-derived components. Besides pluripotent stem cell culture expansion and maintenance, KnockOut™ SR XenoFree CTS™ can be used for hESC⁄hiPSC cryopreservation, derivation, and differentiation studies.
    GIBCO™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
    The KnockOut™ SR XenoFree CTS™ Kit includes a bottle of KnockOut™ SR XenoFree CTS™ as a frozen liquid in a 500mL or a 100 mL PET bottle and a product manual.
  • Specifications
    Specifications
    Stem cells (embryonic), stem cells (iPS - induced pluripotent stem)
    ≤10 EU/mL
    Frozen
    ES cell-qualified
    –20 to –5°C
    –5 to –20°C
    CTS™ KnockOut™ SR XenoFree Kit, 100mL, Frozen, Serum-Free, Xeno-Free, No Glucose, No Glutamine, No HEPES, No Phenol Red indicator, Sterile, Human Species, Used for Clinical and Translational Research, Mammalian Cell Culture, Stem Cell Culture
    Clinical and translational research, mammalian cell culture, stem cell culture
    CTS™ KnockOut™ SR Xeno-Free Kit
    100mL
    HSA
    Sterile
    Safety and Handling
    Safety and Handling
    ShelfLife : 18 Month(s)
    Documents
    Provide Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Cancel Submit
    For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.