Learn More
Gibco™ CTS™ KnockOut™ SR XenoFree Kit Non-distribution product as customer accommodation.
CTS™ KnockOut™ SR XenoFree Kit
$154.00 - $578.00
Specifications
Cell Type Stem Cells (iPS - Induced Pluripotent Stem), Stem Cells (Embryonic)
Sterility Sterile
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A1099201 Non-distribution product as customer accommodation.
View Documents
Gibco™
A1099201
100mL Each for $154.00
Add to cart
A1099202 Non-distribution product as customer accommodation.
View Documents
Gibco™
A1099202
500mL Each for $578.00
Add to cart
Description
Description
GIBCO™ KnockOut™ SR XenoFree CTS™ enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins, which facilitates the transition of hESC and hiPSC research from the bench to the clinic.
The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
  • Defined, xenofree medium for hESC⁄iPSC stem cell culture.
  • First-to-market xenofree hESC⁄iPSC product intended for Ex Vivo tissue and cell culture processing applications.
    GIBCO™ KnockOut™ SR XenoFree CTS™ does not contain bovine or other non-human, animal-derived components. Besides pluripotent stem cell culture expansion and maintenance, KnockOut™ SR XenoFree CTS™ can be used for hESC⁄hiPSC cryopreservation, derivation, and differentiation studies.
    GIBCO™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
    The KnockOut™ SR XenoFree CTS™ Kit includes a bottle of KnockOut™ SR XenoFree CTS™ as a frozen liquid in a 500mL or a 100 mL PET bottle and a product manual.
    “This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
  • Specifications
    Specifications
    Stem Cells (iPS - Induced Pluripotent Stem), Stem Cells (Embryonic)
    Sterile
    For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.