Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTSG Mouse anti-Human, Clone: 9i239, Abnova™

Mouse monoclonal antibody raised against partial recombinant human CTSG.

Manufacturer:  Abnova Corporation MAB12478

Catalog No. 89-934-020


Add to cart

Description

Description

The protein encoded by this gene, a member of the peptidase S1 protein family, is found in azurophil granules of neutrophilic polymorphonuclear leukocytes. The encoded protease has a specificity similar to that of chymotrypsin C, and may participate in the killing and digestion of engulfed pathogens, and in connective tissue remodeling at sites of inflammation. Transcript variants utilizing alternative polyadenylation signals exist for this gene. [provided by RefSeq]

Specifications

Specifications

CTSG
9i239
Mouse monoclonal antibody raised against partial recombinant human CTSG.
In tissue culture supernatant (0.09% sodium azide)
CG/MGC23078
Mouse
IgG1
RUO
Primary
1511
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:50) The optimal working dilution should be determined by the end user.
Liquid
CTSG
Recombinant protein corresponding to 124 amino acids at the N-terminus of human CTSG.
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit