Learn More
CTSS 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00001520T01
Description
Western Blotting
Specifications
Western Blot | |
CTSS 293T Cell Transient Overexpression Lysate(Denatured) | |
36.52 | |
pCMV-CTSS full-length | |
100 μL | |
Human |
293T | |
BC002642 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-CTSS antibody (H00001520-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |