Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTX-II Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259386

Catalog No. NBP259386

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CTX-II
Unconjugated
PBS, pH 7.4 and 50% glycerol with 0.02% Sodium Azide
Cross Linked C-telopeptide of Type II Collagen, CTX2, CTXII
Synthetic peptide corresponding to EKGPDPLQYMRA of human CTXIII, cojugated to OVA..
Immunogen affinity purified
RUO
Primary
Recognizes human Cross Linked C-Telopeptide Of Type II Collagen (CTXII)
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence 5-20 ug/ml, Immunohistochemistry-Frozen 5-20 ug/ml
Affinity Purified
Rabbit
IgG
50 ug
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only