Learn More
Sartorius Cubis™ II Analytical Balance, MCE User Interface
Cubis™ II Analytical Balance features fully customizable hardware, software, and connectivity
Manufacturer:  Sartorius MCE124SUS00
 View more versions of this product
Catalog No. 14-561-261
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor
    (draft shield M), or gesture sensor (optional) learning capability
Specifications
Specifications
Draft shield U - Manual glass
120 g
Cubis™ II Analytical Balance
±0.05 mg
Die-cast Aluminium, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminium Trim
1 sec.
Analytical
120/240 V
2 USB A ports, 1 USB B port, 1USP C port, 1 to 9 pin RS 232 connection, 1 ethernet port
MCE124SUS00
50/60 Hz (±5%)
State of the art TFT touch screen operation with brilliant, readable, display but simple, easy to operate user interface
21 CFR PART 11 17025 USP 41
Balance, Accessories, Power supply, Manual, Factory test report; QP99 App Package
425 mm
100/240 V (±10%)
8.2, 10 kg
2 Years
373 mm
240 mm