Learn More
Sartorius Cubis™ II Analytical Balance, MCE User Interface - PROMO
Cubis™ II Analytical Balance features fully customizable hardware, software, and connectivity
Manufacturer:  Sartorius MCE324SUS00PM
 View more versions of this product
Catalog No. 14-561-271PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor
    (draft shield M), or gesture sensor (optional) learning capability
Specifications
Specifications
Draft shield U - Manual glass
State of the art TFT touch screen operation with brilliant, readable, display but simple, easy to operate user interface
21 CFR PART 11 17025 USP 41
Balance, Accessories, Power supply, Manual, Factory test report; QP99 Software
425 mm
100/240 V (±10%)
8.2, 10.0 kg
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
MCE324P2S00U
240 mm
320 g
50/60 Hz (±5%)
0.1 mg
Die-cast Aluminium, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminium Trim
1 sec.
Analytical
120/240 V
373 mm
± 0.04 mg