Learn More
Sartorius Cubis™ II Micro Balance, MCA User Interface - PROMO
Cubis™ II Micro Balance features fully customizable hardware, software, and connectivity
Specifications
Certifications/Compliance 21 CFR PART 11 17025 USP 41
Display Type 7 in. Color Touch TFT in 16:9 format with new user interface
Frequency 50/60 Hz (±5%)
Type Micro Balance
Voltage 100/240 V (±10%)
View More Specs
Products 4
Catalog Number Mfr. No. Capacity Model Includes Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Model Includes Price Quantity & Availability  
14-561-243PM
View Documents
Sartorius
MCA3.6PMS00PM
1.1 g, 2.1 g, 3.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-253PM
View Documents
Sartorius
MCA6.6SFS00PM
6.1 g Draft shield F: Filtered Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-254PM
View Documents
Sartorius
MCA6.6SMS00PM
6.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-223PM
View Documents
Sartorius
MCA10.6SMS00PM
10.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor
    (draft shield M), or gesture sensor (optional) learning capability
Specifications
Specifications
21 CFR PART 11 17025 USP 41
50/60 Hz (±5%)
100/240 V (±10%)
7 in. Color Touch TFT in 16:9 format with new user interface
Micro Balance