Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Cubis II Precision (1-place) Balance, MCA Interface
Cubis II Precision (1-Place) Balance features fully customizable hardware, software, and connectivity
Manufacturer: Sartorius MCA5202S2S00R
Description
- Monolithical weighing system
- Integrated climate sensors
- Individual sample holders
- Error-free operation
- Individual QApp workflows
- Motorized auto-leveling
- Full end-to-end data integrity
- 21 CFR Part 11 compliance
- Integrated audit-trail
- State-of-the-art user management
- Oustanding service support
- Integrated status center
- Integrated service functions
- Preventive maintenance on accredited standards
- For all models up to a maximum capacity of 8.2 kg
- Large 7-inch color touch TFT display in 16:9 format with new user interface
- Software Factory installed basic set of essential weighing applications (license free) and packages with special weighing applications and function extensions (license required)
- Operation Activated by touch key, touch-free using IR sensor
(draft shield M) or gesture sensor (optional), learning capability
Specifications
Precision (1-place) Balance | |
0.1 g | |
50 to 60 Hz (± 5%) | |
470 mm | |
Die-cast aluminium, plastic PBT, optiwhite float glass and stainless steel, aluminium trim | |
100 to 240 V (± 10%) | |
240 mm | |
5200 g |
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port | |
95 mm | |
Balance, Accessories, Power supply, Manual, Factory test report | |
Draft shield R: Stainless steel frame | |
1 sec | |
5.4kg | |
2 years | |
7 in. color touch TFT display in 16:9 format with new user interface |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title