Learn More
Sartorius Cubis™ II Precision (2-place) Balance, MCE User Interface - PROMO
Cubis™ II Precision (2-Place) Balance features fully customizable hardware, software, and connectivity
Specifications
Certifications/Compliance 21 CFR PART 11 17025 USP 41
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Display Type TFT Touchscreen
Frequency 50/60 Hz (± 5%)
Material Die-cast Aluminium, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminium Trim
View More Specs
Products 4
Catalog Number Mfr. No. Capacity Includes Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Includes Price Quantity & Availability  
14-561-264PM
View Documents
Sartorius
MCE2202S0S00PM
2200 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-270PM
View Documents
Sartorius
MCE32202P0S00PM
4200 g, 32,200 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-274PM
View Documents
Sartorius
MCE4202S0S00PM
4200 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-275PM
View Documents
Sartorius
MCE6202S0S00PM
6200 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor
    (draft shield M), or gesture sensor (optional) learning capability
Specifications
Specifications
21 CFR PART 11 17025 USP 41
TFT Touchscreen
Die-cast Aluminium, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminium Trim
Precision (2-place) Balance
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
50/60 Hz (± 5%)
No draft shield
100/240 V (± 10%)