Learn More
Sartorius Cubis™ II Precision (3-place) Balance, MCA User Interface - PROMO
Cubis™ II Precision (3-Place) Balance features fully customizable hardware, software, and connectivity
Specifications
Certifications/Compliance 21 CFR PART 11 17025 USP 41
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Display Type 7 in. Color Touch TFT in 16:9 Format
Frequency 50/60 Hz (± 5%)
Material Die-cast Aluminium, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminium Trim
View More Specs
Products 5
Catalog Number Mfr. No. Capacity Includes Model Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Includes Model Price Quantity & Availability  
14-561-256PM
View Documents
Sartorius
MCA623SRS00PM
620 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield R: Stainless steel frame This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-230PM
View Documents
Sartorius
MCA1203SRS00PM
1200 g Balance, Accessories, Power supply, Manual, Factory test report. QP99 software Draft shield R: Stainless steel frame This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-240PM
View Documents
Sartorius
MCA2203SRS00PM
2200 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield R: Stainless steel frame This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-245PM
View Documents
Sartorius
MCA3203SRS00PM
3200 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield R: Stainless steel frame This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-561-250PM
View Documents
Sartorius
MCA5203SRS00PM
5200 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield R: Stainless steel frame This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Oustanding service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • For all models up to a maximum capacity of 8.2 kg
  • Large 7-inch color touch TFT display in 16:9 format with new user interface
  • Software Factory installed basic set of essential weighing applications (license free) and packages with special weighing applications and function extensions (license required)
  • Operation Activated by touch key, touch-free using IR sensor
    (draft shield M) or gesture sensor (optional), learning capability
Specifications
Specifications
21 CFR PART 11 17025 USP 41
7 in. Color Touch TFT in 16:9 Format
Die-cast Aluminium, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminium Trim
100/240 V (± 10%)
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
50/60 Hz (± 5%)
Precision (3-place) Balance