Learn More
Sartorius Cubis™ II Precision High-Capacity Balance, MCE User interface - PROMO
Cubis™ II Precision High-Capacity Balance features fully customizable hardware, software, and connectivity
Manufacturer:  Sartorius MCE36200S0S00PM
 View more versions of this product
Catalog No. 14-561-272PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor
    (draft shield M), or gesture sensor (optional) learning capability
Specifications
Specifications
50/60 Hz (± 5%)
High Capacity Precision Balance
21 CFR PART 11 17025 USP 41
126/159 mm
412 mm
No draft shield
1 sec
15.8/17.1 kg
120/240 V
36,200 g
TFT Touchscreen
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
1 g
Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
±0.5
Die-cast Aluminium, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminium Trim
100/240 V (± 10%)
400 mm
2 Years