Learn More
Seca Corp 490 Cuff, Size Medium for seca 535 vital signs monitor
Supplier: Seca Corp 4900026001
A cuff is the perfect sensor for precise measurement results. The perfect fit is decisive for this, which is why the seca 535 mVSA vital signs monitor cuff is available in various sizes. The watertight fabric is easy to disinfect and clean and the entire cuff is latex-free. It was designed for long-term use and made of durable nylon. All cuffs are naturally reusable. • Size M: 24 - 32 cm • For perfect measurement results with seca 535 mVSA vital signs monitor • Anti-allergenic because it is free of latex • Variable cuff sizes from XS to XXL • Reusable cuffs • Nylon material
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.