Learn More
Med Vet International 3.0cm Disposable Blood Pressure Cuff, J0563R, 5 Pack
Supplier: Med Vet International JOR563R
These cuffs are made from a soft, white vinyl. They feature a Velcro closure and will fit all models of blood pressure measurement systems. A single tube. Made in the USA, and inexpensive. White 5-pk. Proper Size is 40% of the circumference of the limb when the cuff is being applied These sizes should cover just about all small animals. Cuff size ranges in centimeters.