Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUL4B Mouse anti-Human, Mouse, Canine, Alexa Fluor™ 700, Clone: OTI1C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271373AF700

Catalog No. N271373A700


Add to cart

Description

Description

CUL4B Monoclonal antibody specifically detects CUL4B in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CUL4B
OTI1C4
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CUL4B
Recombinant protein fragment corresponding to amino acids 1-300 of human CUL4B (NP_003579) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication
Primary
8450.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CUL-4B, cullin 4B, cullin-4B, DKFZp686F1470, KIAA0695, MRXHF2, MRXS15, MRXSC, SFM2
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit