Learn More
R&D Systems™ Cultrex Cell Migration Assay
Standardized assay for the quantification of cell migration
Manufacturer:  R&D Systems™ 3465024K
Catalog No. 34-650-24K
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Originally created in an effort to accelerate the screening process for compounds that influence chemotaxis, which is a fundamental component of angiogenesis, embryonic development, immune responses, and wound healing.
  • Offers a flexible, standardized, multi-well format for quantitating the degree to which cells migrate in vitro in response to chemoattractants in the presence of inhibitors or stimulants.
  • The inserts are sufficiently large to allow for subsequent analysis of migrating subpopulations, in response to chemokines, cytokines, drugs or other compounds of interest.
  • Includes 24 Well Cell Migration Chamber, 25X Cell Wash Buffer, 10X Cell Dissociation Solution 2, and Calcein-AM
Specifications
Specifications
24 Tests