Learn More
R&D Systems™ Cultrex™ Human Fibronectin PathClear™
R&D Systems™ Cultrex™ Human Fibronectin PathClear™ is an extracellular matrix (ECM) component and is one of the primary cell adhesion molecules.
Manufacturer:  R&D Systems™ 342000101
Catalog No. 34-200-0101
Add to cart
This item is not returnable. View return policy
Description
Description
  • The occurrence of different isoforms is due to alternative mRNA splicing of the ED-A, ED-B and III-CS regions, and subsequent post-translational modification.
  • Although non-reactive with adhesion receptors in its soluble state, fibronectin is highly adhesive when immobilized on a surface.
Specifications
Specifications
2335
1mg
Human
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.