Learn More
R&D Systems™ Cultrex™ Stem Cell Qualified Human Vitronectin PathClear™
R&D Systems™ Cultrex™ Stem Cell Qualified Human Vitronectin PathClear™ provides a functionally defined and effective feeder-free surface for the attachment and maintenance of embryonic stem cells in a pluripotent state.
Manufacturer:  R&D Systems™ 342100103
Catalog No. 34-210-0103
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Human plasma
1mg/mL concentration
Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
200μg
Store the unopened product at -70°C