Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cultrex Stem Cell Qualified RGF Basement Membrane Extract

Applications: Bioactivity

$68.44 - $312.04

Specifications

Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Concentration 12 to 18mg/mL
Endotoxin Level ≤ 8EU/mL
For Use With (Application) Bioactivity
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
343400102
View Documents
R&D Systems™
343400102
1 mL
Each for $68.44
Add to Cart
 
343400502
View Documents
R&D Systems™
343400502
5 mL
Each for $312.04
Add to Cart
 
Description

Description

The extract gels at 37°C to form a reconstituted basement membrane. The major components of BME include laminin, collagen IV, entactin, and heparin sulfate proteoglycan. BME can be used in a multiple applications, under a variety of cell culture conditions, for maintaining growth or promoting differentiation of primary endothelial, epithelial, smooth muscle and stem cells. BME can also be utilized in cell attachment, neurite outgrowth, angiogenesis, in vitro cell invasion and in vivo tumorigenicity assays.
Specifications

Specifications

Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
≤ 8EU/mL
Murine Engelbreth-Holm-Swarm (EHS) tumor.
12 to 18mg/mL
Bioactivity
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.