Learn More
Labconco™ Culture Tube Insert Retainer Top
Type 304 stainless steel top for use with Labconco Culture Tube Inserts.
Supplier: Labconco™ 4401401/EMD
Description
- Safely and easily wash and dry culture tubes
- Designed specifically for Labconco culture tube inserts
- Sits on culture tube inserts to ensure safe washing and drying of tubes
- Type 304 stainless steel
- Included with 4588600 (Culture Tube Insert with 240-tube capacity)
- May be placed in upper or lower standard open or spindle rack in Labconco FlaskScrubber or SteamScrubber
Warranty and Services
One year
Specifications
Stainless Steel | |
For Spindle, Standard | |
8.3 in. | |
0.3 in. | |
18.5 in. |
Retainer Top | |
For use with Culture Tube Inserts 4588600, 4402001 and 4402101. | |
21 cm | |
7.6 mm | |
46.9 cm |