Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumene, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
98-82-8
C9H12
120.19
MFCD00008881
RWGFKTVRMDUZSP-UHFFFAOYSA-N
isopropylbenzene,2-phenylpropane,cumol,benzene, 1-methylethyl,1-methylethyl benzene,isopropylbenzol,cumeen,isopropilbenzene,isopropylbenzeen,2-fenilpropano
7406
CHEBI:34656
cumene
CC(C)C1=CC=CC=C1
98-82-8
C9H12
120.19
MFCD00008881
RWGFKTVRMDUZSP-UHFFFAOYSA-N
isopropylbenzene,2-phenylpropane,cumol,benzene, 1-methylethyl,1-methylethyl benzene,isopropylbenzol,cumeen,isopropilbenzene,isopropylbenzeen,2-fenilpropano
7406
CHEBI:34656
cumene
CC(C)C1=CC=CC=C1
Viscosity 0.79 mPa.s (20°C)
Specific Gravity 0.862
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4905 to 1.4925
Melting Point -96.0°C
Color Colorless
Density 0.8620g/mL
Boiling Point 152.0°C to 154.0°C
Quantity 1L
Flash Point 31°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5CH(CH3)2
Beilstein 05, 393
Merck Index 15, 2607
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol and many other organic solvents,carbon tetrachloride,ether,benzene,acetone
Formula Weight 120.19
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material Cumene, 99%
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortabl

GHS Signal Word: Danger

EINECSNumber : 202-704-5

RTECSNumber : GR8575000

TSCA : TSCA