Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumene Hydroperoxide, 77%, Spectrum™

Click to view available options
Quantity:
25 mL
100 mL
500 mL
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
80-15-9
C9H12O2
152.19
YQHLDYVWEZKEOX-UHFFFAOYSA-N
2-phenylpropane-2-peroxol
CC(C)(OO)C1=CC=CC=C1
80-15-9
C9H12O2
152.19
YQHLDYVWEZKEOX-UHFFFAOYSA-N
2-phenylpropane-2-peroxol
CC(C)(OO)C1=CC=CC=C1

C2313, 80-15-9, C9H12O2

Spectrum™ Cumene Hydroperoxide , also known as Cumyl Hydroperoxide, is used as an oxidizing agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Poly Bottle
Quantity 100 mL
CAS Max % 1
UN Number UN3109
Percent Purity 77%