Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumyl hydroperoxide, 80%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
2.5kg
Packaging:
Plastic bottle
80-15-9,98-82-8,617-94-7,98-86-2
C9H12O2
152.19
MFCD00002129
YQHLDYVWEZKEOX-UHFFFAOYSA-N
cumene hydroperoxide,cumyl hydroperoxide,hydroperoxide, 1-methyl-1-phenylethyl,cumenyl hydroperoxide,alpha,alpha-dimethylbenzyl hydroperoxide,cumolhydroperoxid,7-cumyl hydroperoxide,cumolhydroperoxide,cument hydroperoxide,hydroperoxyde de cumene
6629
CHEBI:78673
2-hydroperoxypropan-2-ylbenzene
CC(C)(C1=CC=CC=C1)OO
80-15-9,98-82-8,617-94-7,98-86-2
C9H12O2
152.19
MFCD00002129
YQHLDYVWEZKEOX-UHFFFAOYSA-N
cumene hydroperoxide,cumyl hydroperoxide,hydroperoxide, 1-methyl-1-phenylethyl,cumenyl hydroperoxide,alpha,alpha-dimethylbenzyl hydroperoxide,cumolhydroperoxid,7-cumyl hydroperoxide,cumolhydroperoxide,cument hydroperoxide,hydroperoxyde de cumene
6629
CHEBI:78673
2-hydroperoxypropan-2-ylbenzene
CC(C)(C1=CC=CC=C1)OO
Oxidation reagent
Specific Gravity 1.06
Density 1.0600g/mL
Melting Point -30.0°C
Flash Point 83°C
Packaging Plastic bottle
Quantity 250g
Assay Percent Range 80 to 85% (Iodometry)
Linear Formula C6H5C(CH3)2OOH
Solubility Information Solubility in water: miscible.
Formula Weight 152.19
Percent Purity 80%
Chemical Name or Material Cumyl hydroperoxide, 80%
Show More Show Less

GHS H Statement
Heating may cause a fire.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause re

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Danger

EINECSNumber : 201-254-7

RTECSNumber : MX2450000

TSCA : TSCA