Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Acetate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
6046-93-1
C4H8CuO5
199.649
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate,cupric acetate monohydrate,copper diacetate monohydrate,copper acetate monohydrate,copper 2+ acetate, monohydrate,acetic acid, copper 2+ salt, monohydrate,verdigris,copper acetate hydrate,diacetoxycopper hydrate,copper acetate, hydrate
165397
copper;diacetate;hydrate
CC(=O)[O-].CC(=O)[O-].O.[Cu+2]
6046-93-1
C4H8CuO5
199.649
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate,cupric acetate monohydrate,copper diacetate monohydrate,copper acetate monohydrate,copper 2+ acetate, monohydrate,acetic acid, copper 2+ salt, monohydrate,verdigris,copper acetate hydrate,diacetoxycopper hydrate,copper acetate, hydrate
165397
copper;diacetate;hydrate
CC(=O)[O-].CC(=O)[O-].O.[Cu+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 199.65
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.88kg
Quantity 2.5kg
Chemical Name or Material Cupric Acetate, Monohydrate