Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Acetate TS, Stronger, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle

C-304, 6046-93-1

Spectrum™ Cupric Acetate TS, Stronger, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 6046-93-1,7732-18-5
CAS Max % 0.001,0.999
Molecular Formula C4H8CuO5
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O
IUPAC Name copper(2+) diacetate hydrate
Molecular Weight (g/mol) 199.65
Packaging Amber Glass Bottle
Quantity 100 mL