Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Acetate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
6046-93-1
C4H8CuO5
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper(2+) diacetate hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O
6046-93-1
C4H8CuO5
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper(2+) diacetate hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O

C-301, 6046-93-1

Spectrum™ Cupric Acetate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L