Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Bromide, Crystal, 98%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7789-45-9
7789-45-9
Grade Reagent
CAS Max % 1
Percent Purity 0.98
Chloride 0.003
Insoluble Matter 0.0001
Iron (Fe) 5e-05
Packaging Amber Glass Bottle
Sulfate 0.0001
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.