Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Carbonate Basic, Powder, 53%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
12069-69-1
12069-69-1
CAS Max % 1
Percent Purity 0.53
Grade Reagent
Chloride 0.0001
Iron (Fe) 0.0001
Packaging Poly Bottle
Sulfate 0.0001
Quantity 125 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.