Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10125-13-0
10125-13-0
Grade ACS
CAS Max % 1
Percent Purity 0.99
Calcium (Ca) 5e-05
Insoluble Matter 0.0001
Iron (Fe) 5e-05
Nickel (Ni) 0.0001
Nitrate 0.00015
Packaging Poly Pail
Potassium (K) 0.0001
Sodium (Na) 0.0002
Sulfate 5e-05
Quantity 12 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.