Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Chloride, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

10125-13-0
Cl2CuH4O2
170.476
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
61482
CHEBI:86318

High quality chemicals for laboratory and specialized industrial use

SMILES O.O.Cl[Cu]Cl
IUPAC Name dichlorocopper;dihydrate
Formula Weight 170.48
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.54kg
Quantity 2.5kg
Chemical Name or Material Cupric Chloride, Dihydrate
Grade Baker Analyzed™ ACS Reagent