Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Chloride, Dihydrate, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10125-13-0
Cl2CuH4O2
170.48
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
copper(2+) dihydrate dichloride
O.O.[Cl-].[Cl-].[Cu++]
10125-13-0
Cl2CuH4O2
170.48
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
copper(2+) dihydrate dichloride
O.O.[Cl-].[Cl-].[Cu++]

C1018, 10125-13-0, CuCl2•2H2O

Spectrum™ Cupric Chloride, Dihydrate, USP - All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
UN Number UN2802
Percent Purity 99 to 100.5%
Calcium (Ca) 5e-005
Insoluble Matter 0.0001
Iron (Fe) 5e-005
Loss on Drying 20.9-21.4%
Nickel (Ni) 0.0001
Packaging Poly Pail
Potassium (K) 0.0001
Sodium (Na) 0.0002
Sulfate 5e-005
Quantity 12 kg
Show More Show Less