Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Hydroxide, Technical, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Amber Glass Bottle
20427-59-2
20427-59-2
Grade Technical
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 2.5 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.