Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Nitrate, Hydrate, Crystal, Purified, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
19004-19-4
Cu2H10N4O17
465.18
IZDJJEMZQZQQQQ-UHFFFAOYSA-N
dicopper(2+) pentahydrate tetranitrate
O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
19004-19-4
Cu2H10N4O17
465.18
IZDJJEMZQZQQQQ-UHFFFAOYSA-N
dicopper(2+) pentahydrate tetranitrate
O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

C1407, 19004-19-4, Cu(NO3)2•2.5H2O

Spectrum™ Cupric Nitrate, Hydrate, Crystal, Purified is a also known as a copper nitrate and can be obtained as a crystalline solid as well as a hydrated form. Mostly used to produce copper oxide, it is also used as a polisher for other metals and in pyrotechnics.
CAS Max % 100%
UN Number UN1477
Packaging Poly Pail
Quantity 12 kg