Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Oxide, Black, Powder, CP, 96%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1317-38-0
1317-38-0
Grade Chemically Pure
CAS Max % 1
Percent Purity 0.96
Lead (Pb) 0.0001
Packaging Fiber Drum
Quantity 45 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.