Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, 0.016 M Solution, Spectrum™

Click to view available options
Quantity:
1 gal
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 7758-99-8,7732-18-5
Concentration 0.01602 ± 0.0002 M
Packaging Poly Bottle
Quantity 1 gal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.