Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cupric Sulfate, 5-Hydrate, U.S.P., J.T.Baker™
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Formula Weight | 249.68 |
Grade | USP |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 2.28kg |
Quantity | 500g |
Chemical Name or Material | Cupric Sulfate, 5-Hydrate |