Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, 5-Hydrate, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
7758-99-8
CuH10O9S
249.677
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
7758-99-8
CuH10O9S
249.677
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
Formula Weight 249.68
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.28kg
Quantity 2.5kg
Grade USP
Chemical Name or Material Cupric Sulfate, 5-Hydrate