Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Anhydrous, Powder, 98%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7758-98-7
7758-98-7
Grade Reagent
CAS Max % 1
Percent Purity 0.98
Lead (Pb) 5e-05
Loss on Drying 0.01
Packaging Poly Bottle
Quantity 125 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.