Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cupric Sulfate, Colorimetric Solution (CS), 1 mL = 62.4 mg CuSO4.5H2O, 61.9-62.9 mg/mL, Spectrum™ Chemical
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Cupric Sulfate, Colorimetric Solution (CS), 1 ml = 62.4 mg CuSO4.5H2O , also known as Copper(II) Sulfate, is an aqueous solution that contains 62.4 mg CuSO4.5H2O per ml.
Specifications
CAS Max % | 6.5%, 92.5% |
UN Number | UN1789 |
Concentration | 61.9-62.9 mg/mL |
Grade | Chemical Solution |
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction