Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Fine Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
2.5kg
Packaging:
Poly Bottle
7758-99-8
CuH10O9S
249.677
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
7758-99-8
CuH10O9S
249.677
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 249.68
Physical Form Fine Crystal
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.28kg
Quantity 500g
Chemical Name or Material Cupric Sulfate