Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Large Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7758-99-8
CuH10O9S
249.677
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
CHEBI:31440

High quality chemicals for laboratory and specialized industrial use

CAS 7758-99-8
Molecular Formula CuH10O9S
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
IUPAC Name copper;sulfate;pentahydrate
Molecular Weight (g/mol) 249.677
PubChem CID 24463
ChEBI CHEBI:31440
Formula Weight 249.68
Physical Form Large Crystal
Packaging Poly Bottle
Specific Gravity 1L = 2.28kg
Quantity 500g
Chemical Name or Material Cupric Sulfate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less