Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Large Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7758-99-8
CuH10O9S
249.677
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 249.68
Physical Form Large Crystal
Packaging Poly Bottle
Specific Gravity 1L = 2.28kg
Quantity 2.5kg
Chemical Name or Material Cupric Sulfate
Grade Baker Analyzed™ ACS Reagent