Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Monohydrate, Purified, 34%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10257-54-2
10257-54-2
Grade Purified
CAS Max % 1
Percent Purity 0.34
Lead (Pb) 0.0001
Nickel (Ni) 5e-05
Packaging Poly Bottle
Quantity 2.5 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.