Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Pentahydrate, BiotechGrade, 98.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7758-99-8
CuH10O9S
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
7758-99-8
CuH10O9S
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

C1421, 7758-99-8, CuSO4•5H2O

Spectrum™ Cupric Sulfate, Pentahydrate, BiotechGrade also known as Copper(II) Sulfate is a chemical compound that exists as a salt or crystal and differs in the degree of hydration. It uses very from use as a herbicide to use as a dye. It is also used in Benedict′s solution, Fehling′s solution and Biuret reagent. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
CAS Max % 1
Appearance Blue, crystallin
UN Number UN3077
Percent Purity 98.5 to 100.5%
Loss on Drying 33.0-36.5%
Packaging Poly Bottle
Quantity 2.5 kg