Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Pentahydrate, Fine Crystal, ACS, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7758-99-8
7758-99-8
Grade ACS
CAS Max % 1
Percent Purity 98 to 102%
Calcium (Ca) 5e-05
Chloride 1e-05
Insoluble Matter 5e-05
Iron (Fe) 3e-05
Nickel (Ni) 5e-05
Nitrogen Compounds 2e-05
Packaging Poly Pail
Potassium (K) 0.0001
Sodium (Na) 0.0002
Quantity 12 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.