Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cupric Sulfate Pentahydrate (Granular/USP), Fisher Chemical
Click to view available options
Poly Bottle; 500g
Specifications
Melting Point | 110°C |
Color | Blue |
CAS | 7758-99-8 |
pH | 3.5 to 4.5 |
Physical Form | Solid |
Quantity | 500 g |
Assay Percent Range | (Dried Basis) 98.5to100.5% |
Molecular Formula | CuH10O9S |
MDL Number | MFCD00149681 |
Synonym | copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas |
Show More |
Safety and Handling
Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. NFPA Health:2 Flammability:0 Instability:1 |
DOTInformation | DOT Class 9, : Miscellaneous |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction